EP Reference Solution BY4, Brownish-Yellow Reference Solution
EP Reference solution BY4, brownish-yellow reference solution, Glass bottle, amber, 120 mL
Catalog # RICC28744
Supplier: RICCA CHEMICAL
EP Reference Solution BY4, Brownish-Yellow Reference Solution
Catalog # RICC28744
Supplier: RICCA CHEMICAL
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle, amber
- Pk:120 mL
- Description:EP Reference solution BY4, brownish-yellow reference solution
- Cat. No.:RICC28744
- Storage temperature:Ambient
- Appearance:Brownish-yellow liquid
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



