ContactUsLinkComponent
M-Enterococcus Agar Base Dehydrated Culture Media, HiMedia
M-Enterococcus Agar Base Dehydrated Culture Media, HiMedia
Catalog # HIMEM1108-500G
Supplier:  HIMEDIA
New Product
CAS Number:  
M-Enterococcus Agar Base Dehydrated Culture Media, HiMedia
Catalog # HIMEM1108-500G
Supplier:  HIMEDIA
New Product
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    M-Enterococcus agar base, 500 g
Specifications
About this item
For the isolation and enumeration of enterococci in water, sewage, food and other materials by membrane filter technique as well as direct plating of specimens.
  • Microbiological culture media
  • 500 g poly bottles