Normal Human Serum
Bottle
Catalog # GEMB100-110-100
Supplier: GEMINI BIO-PRODUCTS
Jump to:
Product Details & Documents
Normal human serum is collected from healthy male and female donors of all ABO serotypes at FDA-licensed facilities in the United States. This material is defibrinated from source plasma. All donor units are tested for viral markers and found to be non-reactive.
- 0,1 µm sterile-filtered
Delivery information: Ships on dry ice.
Specifications
- Pack type:Bottle
- Origin:US
- Cat. No.:GEMB100-110-100
- Storage temperature:Store frozen at −20...−10 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



