ContactUsLinkComponent
TASKI® Jontec Eternum High Gloss Floor Emulsion
TASKI® Jontec Eternum High Gloss Floor Emulsion
Catalog # DIVY7512694
Supplier:  Diversey
New Product
TASKI® Jontec Eternum High Gloss Floor Emulsion
Catalog # DIVY7512694
Supplier:  Diversey
New Product
Specifications
  • Capacity:
    2×5 L
  • Description:
    TASKI Jontec Eternum F2e, high gloss floor emulsion
  • Pack type:
    Box
  • Pk:
    2 Can
Specifications
About this item
High-gloss, slip resistant floor emulsion for most types of smooth and water resistant, hard floors. Its technology provides superior traffic resistance against scuffs and dirt. It ensures reduced maintenance costs to improve productivity. It will be ideal in all prestige and heavy traffic areas where ordinary emulsions require a high level of maintenance.
  • High-gloss, protective layer
  • Hard, durable and wear resistant protection
  • Excellent adhesion to most floor types
  • Slip resistant according to international standards (ASTM D-2047)
  • Easy to repair
Suitable for application with traditional methods as well as with TASKI ProSpeed. TASKI ProSpeed is an easy to use, ergonomic floor finish applicator that will provide a superior quality of the floor finish layer and improve the productivity of applying the floor finish while reducing chemical consumption and packaging waste. Through TASKI ProSpeed, the product consumption can be controlled while providing a uniform top quality layer of floor finish.
Benefits: Enhances the natural appearance of the floor by providing a long lasting high gloss finish. Superior resistance to scuff marks and dirt, even under heavy traffic. Suitable for most types of floors such as vinyl, synthetic, linoleum, stone, sealed wood and cork floors. Spreads evenly for perfect levelling. Guaranteed slip fall prevention and safety for your employees and customers. Responds to most common maintenance methods. Suitable for traditional applications with a flat mop, mop and bucket or standard polish applicators. To ensure superior results, choose the application with the ProSpeed floor finish applicator tool which is proven to reduce the operator’s fatigue against traditional application methods while improving results and productivity.