Vasco® OP eco Surgical and Protective Gloves, Sterile
Supplier: B. Braun
New Product
Certificates
About this item
Protection against particulate radioactive contamination.
Protein content ≤ 50 μg/g, lower claims are not considered to be reliable given the expected process variation in manufacture and inter-laboratory testing (EN 455-3).
Contains natural rubber latex which may cause allergic reactions including anaphylactic reactions.
Specifications
- Catalog No:
- BRAU6081334
- BRAU6081332
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Latex / latex-free:
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Material:
- Natural rubber latex
- Natural rubber latex
- Cuff style:
- Rolled
- Rolled
- Palm thickness:
- —
- 89 ± 5 mm
- Sterility:
- Sterile
- Sterile