Order Entry
Switzerland
ContactUsLinkComponent
MP650 Dye qPCR Calibration Solution *10000X*
MP650 Dye qPCR Calibration Solution *10000X*
Catalog # AATB67039
Supplier:  AAT Bioquest
New Product
CAS Number:  
undefined
MP650 Dye qPCR Calibration Solution *10000X*
Catalog # AATB67039
Supplier:  AAT Bioquest
New Product
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Description:
    MP650 Dye qPCR calibration solution *10000X*

Specifications

About this item

MP650 is designed to have identical spectra to those of Mustang Purple.

  • Stable for 12 months

Quality tested in-house.

Caution: For research use only.