iFluor® 750-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # AATB25562
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- VE:1 mg
- Bezeichnung:iFluor® 750-Wheat Germ Agglutinin (WGA) conjugate
Specifications
About this item
iFluor® 750 labelled WGA exhibits a distinct far-red fluorescence characteristic, emitting maximally at 779 nm.
- Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.