ContactUsLinkComponent
Buccutite™ Rapid APC-Cy7 Tandem Antibody Labelling Kit
Buccutite™ Rapid APC-Cy7 Tandem Antibody Labelling Kit
  AATB5415
New Product
 :  
Buccutite™ Rapid APC-Cy7 Tandem Antibody Labelling Kit
  AATB5415
New Product
 :  
  • Pk:
    2
  • Includes:
    2 labellings
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ rapid APC-Cy7 tandem antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PE, APC, PerCP and iFluor® tandem dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
 : Shipped with blue ice.
 : Research use only.