ContactUsLinkComponent
Buccutite™ Rapid PE-Cy7 Tandem Antibody Labelling Kit
Buccutite™ Rapid PE-Cy7 Tandem Antibody Labelling Kit
Catalog # AATB5411
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Buccutite™ Rapid PE-Cy7 Tandem Antibody Labelling Kit
Catalog # AATB5411
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    2
  • Includes:
    2 labellings
Specifications
About this item
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ Rapid PE-Cy7 tandem antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PE, APC, PerCP and iFluor® tandem dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped with blue ice.
Caution: Research use only.