ContactUsLinkComponent
Portelite™ Fluorimetric Lithium Ion Quantification Kit
Portelite™ Fluorimetric Lithium Ion Quantification Kit
Catalog # AATB21353
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Portelite™ Fluorimetric Lithium Ion Quantification Kit
Catalog # AATB21353
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    50 Tests
  • Description:
    Portelite™ Fluorimetric Lithium Ion Quantification Kit
Specifications
About this item
Designed for consistent and dependable results. Portelite™ fluorimetric lithium ion quantification kit uses our robust lithium ion indicator dye, Lithiumighty™ 520, which exhibit great fluorescence intensity enhancement upon binding to lithium ions.
  • Lithiumighty ™ 520 is perhaps the most robust lithium-ion indicator with high selectivity
  • Rapid determination of lithium concentrations in a variety of samples
  • 12-month shelf life
This nanodrop-based assay kit requires a small amount of sample, it is particularly suitable for the determination of lithium ion concentration in fields or on site.
Delivery: Shipped at RT.
Caution: Research use only.