ReadiLink™ Rapid trFluor™ Tb Antibody Labelling Kit
Catalog # AATB1305
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:2
- Includes:2 labellings
Specifications
About this item
Microscale optimised for labelling 50 µg antibody per reaction. Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality labelled antibodies for multiple downstream applications.
Delivery: Shipped at RT.
Caution: Research use only.