Order Entry
Switzerland
ContactUsLinkComponent
Amplite® Fluorimetric DPP4 Activity Assay Kit
Amplite® Fluorimetric DPP4 Activity Assay Kit
Catalog # AATB11323
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Amplite® Fluorimetric DPP4 Activity Assay Kit
Catalog # AATB11323
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pk:
    100 Tests

Specifications

About this item

Designed for consistent and dependable results. The Amplite® fluorimetric DPP4 activity assay kit provides a simple, quick, and direct protocol for measuring DPP4 Activity.

  • Ex/Em = 360/460 nm
  • Assay is compatible with high-throughput screening systems

The fluorescence intensity generated is directly proportional to the DPP4 activity in the sample, making it suitable for detecting DPP4 in various biological matrices such as cellular lysates, tissue extracts and serum.

Delivery: Shipped with blue ice.
Caution: Research use only.