Order Entry
Switzerland
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Pullulan Standard
  18579-25MG
 :  Merck
 :  
Supelco® Organic Reference Standard, Pullulan Standard
  18579-25MG
 :  Merck
 :  

 

  • Pk:
    25 mg
  • Standards:
    Certified reference material, certified according to DIN, for GPC, 400000
  • Description:
    Pullulan Standard
  • Seq:
    22
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Flash point:
    Not applicable