Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Pullulan Standard
Supelco® Organic Reference Standard, Pullulan Standard
Catalog # 18179-25MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Pullulan Standard
Catalog # 18179-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Certified reference material, certified according to DIN, for GPC, 6000
  • Description:
    Pullulan Standard
  • Seq:
    12
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Flash point:
    Not applicable

Specifications

About this item