Buccutite™ Rapid PE Antibody Labelling Kit
AATB5405
New Product
:
- Pk:2
- Includes:2 labellings
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ Rapid PE antibody labelling kits, designed for large-scale production, offer a convenient and efficient method to label antibodies with PE, APC and iFluor® tandem dyes.
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
: Shipped with blue ice.
: Research use only.