Supelco® Organic Reference Standard, L-Rhamnose Monohydrate
41651-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:10030-85-0
- Description:ʟ-Rhamnose monohydrate
- Storage conditions:No temperature limit
- Shipping conditions:STD
- MDL No.:MFCD00071591
- M-Clarity quality level:MQ100
- Synonym(s):6-Deoxy-ʟ-mannose; ʟ(+)Rhamnopyranose
- Molecular weight:182,17