Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Tigloyl-L-Carnitine-(N-Methyl-d₃)
Supelco® Organic Reference Standard, Tigloyl-L-Carnitine-(N-Methyl-d₃)
Catalog # 12633-1MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Tigloyl-L-Carnitine-(N-Methyl-d₃)
Catalog # 12633-1MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Standards:
    Analytical standard
  • Description:
    Tigloyl-ʟ-carnitine-(N-methyl-d₃)
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    WICE
  • Synonym(s):
    (2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d₃)-2-[[(2E)-2-methyl-1-oxo-2-butenyl]oxy]-1-propanaminium inner salt
  • Molecular weight:
    246,32

Specifications

About this item