Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Valeryl-L-Carnitine-(N-Methyl-d₃)
Supelco® Organic Reference Standard, Valeryl-L-Carnitine-(N-Methyl-d₃)
Supplier:  Merck
Certificates
Supelco® Organic Reference Standard, Valeryl-L-Carnitine-(N-Methyl-d₃)
Supplier:  Merck

Specifications

  • Storage conditions:
    –20 °C
  • Shipping conditions:
    WICE
  • Synonym(s):
    (2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d₃)-2-[(1-oxopentyl)oxy]-1-propanaminium inner salt
  • Molecular weight:
    248,33

Specifications

Frequently Bought Together