ReadiView™ Blue Real-Time PCR Visualisation Dye *200X*
AATB17300
New Product
:
- Pk:500 Tests
- Description:ReadiView™ blue real-time PCR visualisation dye *200X*
ReadiView™ Blue Real-Time PCR Visualisation Dye is a 200X concentrated, ready-to-use, inert dye that enhances the visibility of real-time PCR reactions for more accurate pipetting and plate loading.
- Stable for 12 months
Quality tested in-house.
: For research use only.