ReadiPrep™ Nickel NTA Magnetic Beads
Supplier: AAT BIOQUEST
New Product
Certificates
About this item
ReadiPrep™ Nickel NTA magnetic beads provide a fast and simple solution for small-scale affinity purification of histidine-tagged proteins.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together