ContactUsLinkComponent
mFluor™ Violet 450-Wheat Germ Agglutinin (WGA) Conjugate
mFluor™ Violet 450-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # AATB25450
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
mFluor™ Violet 450-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # AATB25450
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    1 mg
  • Description:
    mFluor™ Violet 450-Wheat germ agglutinin (WGA) conjugate
Specifications
About this item
mFluor™ Violet 450 labelled WGA is well-excited by the violet laser, emitting a bright blue fluorescence at 445 nm.
  • Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.