ContactUsLinkComponent
iFluor® 700-Wheat Germ Agglutinin (WGA) Conjugate
iFluor® 700-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # AATB25561
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
iFluor® 700-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # AATB25561
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    1 mg
  • Description:
    iFluor® 700-Wheat germ agglutinin (WGA) conjugate
Specifications
About this item
The iFluor® 700 conjugate of WGA possesses a bright, far-red fluorescence characteristic of the iFluor® 700 dye (Ex/Em = 690/713 nm).
  • Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.