ContactUsLinkComponent
HIS Lite™ OG488-Tris NTA Chelator
HIS Lite™ OG488-Tris NTA Chelator
  AATB12655
New Product
 :  
HIS Lite™ OG488-Tris NTA Chelator
  AATB12655
New Product
 :  
  • Pk:
    100 µG
  • Description:
    HIS Lite™ OG488-tris NTA chelator
HIS Lite™ OG488-Tris NTA Chelator is used as a sensitive green fluorescent probe for detecting polyhistidine-labelled proteins in combination with the addition of a heavy metal ion such as Ni²⁺ in cells, solution and solid surfaces.
  • Stable for 24 months
Quality tested in-house.
 : For research use only.