HIS Lite™ Cy3 Tris NTA Chelator
Catalog # AATB12660
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:100 µG
- Description:HIS Lite™ Cy3 tris NTA chelator
Specifications
About this item
Cy3-Tris NTA compound is a sensitive fluorescent chelator for detecting polyhistidine-labelled proteins in cells, solution, and solid surfaces with the addition of certain metal ions (such as Ni²⁺, Co₂⁺ and more) as a mediator.
- Stable for 24 months
Quality tested in-house.
Caution: For research use only.