Order Entry
Switzerland
ContactUsLinkComponent
 
Bole white (kaolin), GPR RECTAPUR® , washed
New Product
 
undefined
Bole white (kaolin), GPR RECTAPUR® , washed
New Product
 

 

  • Identity:
    Passes Test
  • Substances soluble in HCl:
    max. 1.0%
  • Pb (Lead):
    max. 0.001%
  • Loss on ignition (600°C):
    max. 15.0%
  • Water :
    max. 1.5%

 

Frequently Bought Together