Supelco® Organic Reference Standard, Oil and Grease 1000 mg/L Calibration Standard
Catalog # OG1000-250ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:In H₂O
- Pk:250 mL
- Standards:Certified reference material
- Description:Oil and grease 1000 mg/L calibration standard
- Storage conditions:2...30 °C
- Hazard class description:H318
- Shipping conditions:STD
- M-Clarity quality level:MQ100
Specifications
About this item
- GHS signal: GHS05
Caution: P280 - P305 + P351 + P338