Order Entry
Switzerland
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Ethiofencarb
Catalog # CRM05286-100MG
Anbieter:  Merck
CAS-Nummer:  
Supelco® TraceCERT® Organic Reference Standard, Ethiofencarb
Catalog # CRM05286-100MG
Anbieter:  Merck
CAS-Nummer:  

Technische Daten

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    29973-13-5
  • Description:
    Ethiofencarb
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302 - H410
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00055463
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    225,31

Technische Daten

Über diesen Artikel

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The CRM may also find the following uses:

  • Estimation of systemic and non-systemic pesticides in 80 samples of apples from Kazakhstan by LC-MS/MS and GC-MS/MS
  • Development of a method for rapid multi-residue screening of 12 pesticides and 7 mycotoxins in grain and animal feed samples using flow injection (FI) mass spectrometry (MS)
  • UHPLC-MS/MS determination of carbamate pesticide residues in fresh herb samples for human consumption and an optimised QuEChERS extraction
  • Study the photocatalytic degradation of ethiofencarb in the presence of a chalcogenide, SnI₄S₈
  • Differentiation between structurally isomeric pesticides using high-resolution accurate mass analyzers
  • Multi-residue analysis of 250 pesticides by ultra-performance liquid chromatography (UHPLC) coupled to MS/MS in processed fruits
Certifications: The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The CRM may also find the following uses:
  • Estimation of systemic and non-systemic pesticides in 80 samples of apples from Kazakhstan by LC-MS/MS and GC-MS/MS
  • Development of a method for rapid multi-residue screening of 12 pesticides and 7 mycotoxins in grain and animal feed samples using flow injection (FI) mass spectrometry (MS)
  • UHPLC-MS/MS determination of carbamate pesticide residues in fresh herb samples for human consumption and an optimized QuEChERS extraction
  • Study the photocatalytic degradation of ethiofencarb in the presence of a chalcogenide SnI4S8
  • Differentiation between structurally isomeric pesticides using high-resolution accurate mass analyzers
  • Multi-residue analysis of 250 pesticides by ultra-performance liquid chromatography (UHPLC) coupled to MS/MS in processed fruits
Caution: P264 - P270 - P273 - P301 + P312 - P391 - P501