Supelco® TraceCERT® Organic Reference Standard, Dichlon
N° de catalogue CRM04144-100MG
Fournisseur: Merck
Numéro CAS:
Spécifications
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:117-80-6
- Description:Dichlon
- Storage conditions:15...25 °C
- Hazard class description:H301 - H315 - H319 - H410
- Shipping conditions:STD
- MDL No.:MFCD00001677
- M-Clarity quality level:MQ300
- Synonym(s):2,3-Dichloro-1,4-naphthoquinone; Dichlon
- Melting point:194 - 197 °C (lit.)
- Molecular weight:227,04
Spécifications
À propos de cet article
It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Dichlone CRM may also find its use as mentioned below:
- To evaluate the toxicity of some naturally occurring quinones and easily synthesised quinone derivatives on the two-spotted spider mite and three aphid species in laboratory and greenhouse conditions
- To assess antifeedant and toxic effects of selected quinones against the cabbage looper, Trichoplusia ni larvae
- Evaluation of four 1,4-naphthoquinones and 10 pyrimidinone-fused 1,4-naphthoquinones for antimicrobial activity against drug-resistant oral bacteria
- To determine the effectiveness of the use of Supercritical Fluid Extraction (SFE) and Supercritical Fluid Chromatography (SFC) techniques in the analysis of the levels of pesticide residues in canned foods, vegetables, and fruits
Attention: P264 - P270 - P273 - P301 + P310 - P302 + P352 - P305 + P351 + P338