Supelco® TraceCERT® Organic Reference Standard, Teflubenzuron
CRM01399-100MG
: Merck
:
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:83121-18-0
- Description:Teflubenzuron
- Storage conditions:2…8 °C
- Hazard class description:H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD00068254
- M-Clarity quality level:MQ300
- Synonym(s):Clofentezine
- Molecular weight:381,11
It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. The teflubenzuron certified reference material (CRM) may also find use as given below:
- Determination of 9 benzoylurea insecticides in fruit vegetable cereal and animal product samples by pressurised liquid extraction and LC-MS/MS
- Simultaneous analysis of lufenuron bifenthrin and teflubenzuron residues in coconut water by matrix solid-phase dispersion extraction combined with reversed-phase high-performance liquid chromatography
- Development of a high performance liquid chromatography-triple quadrupole mass spectrometry (HPLC-MS/MS) based method for the simultaneous estimation of 7 benzoylurea insecticides in infused oolong tea
- Multi-residue analysis of pesticides in Açai fruit by an extraction based on matrix solid phase dispersion and determination using liquid chromatography
- Comparative study for the extraction of 9 benzoylurea insecticides using two dispersive liquid-liquid microextraction (DLLME) approaches from wastewater samples followed by their determination using liquid chromatography-quadrupole-linear ion trap-mass spectrometry
: P260 - P273 - P314 - P391 - P501