Order Entry
Switzerland
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, Fluopyram
  CRM01345-25MG
 :  Merck
 :  
Supelco® TraceCERT® Organic Reference Standard, Fluopyram
  CRM01345-25MG
 :  Merck
 :  

 

  • Pk:
    25 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    658066-35-4
  • Description:
    Fluopyram
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H411
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD17166960
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    N-[2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]ethyl]-2-(trifluoromethyl)benzamide; N-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]ethyl}-α,α,α-trifluoro-o-toluamide
  • Molecular weight:
    396,71

 

 

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Fluopyram CRM may also find its use as described below:

  • Method development for analyzing the succinate-dehydrogenase-inhibitor fungicide residue levels in vegetable and fruit samples by liquid chromatography-tandem mass spectrometry
  • Development and validation of monoclonal antibody-based immunoassays to analyze fluopyram residues in food samples
  • Simultaneous estimation of fluopyram and tebuconazole in watermelon and soil samples using QuEChERS and GC-MS based method
  • Determination of fluopyram flupyradifurone and indaziflam in date fruit pistachio and soil samples by ultra-high performance liquid chromatography with tandem mass spectrometry(UHPLC-MS)
  • Develop and validate a method based on QuEChERS and UHPLC-Orbitrap-MS to determine fluopyram and its metabolites in cherry tomato and cucumber
 : P273 - P391 - P501