Supelco® Organic Reference Standard, Bicyclohexyl
Catalog # 94057-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:92-51-3
- Description:Bicyclohexyl
- Environmentally Preferable:
- Storage conditions:No temperature limit
- Hazard class description:H302 - H304 - H335 - H410
- Shipping conditions:STD
- MDL No.:MFCD00003815
- M-Clarity quality level:MQ100
- Synonym(s):Dicyclohexyl
- Melting point:3 - 4 °C (lit.)
- Formula:C₆H₁₁C₆H₁₁
- Molecular weight:166,30
- Density:0,864 g/ml at 25 °C (lit.)
- Flash point:92 °C - Closed cup
- Boiling point:227 °C (lit.)
Specifications
About this item
Caution: P261 - P264 - P270 - P273 - P301 + P310 - P331