Supelco® TraceCERT® Organic Reference Standard, Bromopropylate
Catalog # 93744-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:18181-80-1
- Description:Bromopropylate
- Storage conditions:15...25 °C
- Hazard class description:H410
- MDL No.:MFCD00078632
- M-Clarity quality level:MQ300
- Synonym(s):Isopropyl 4,4′-dibromobenzilate; Phenisobromolate
- Molecular weight:428,12
Specifications
About this item
The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Bromopropylate CRM may also find following uses:
- To elucidate the inhibition of plasma and brain acetylcholinesterase activity in Wistar rats with a mixture of 5 pesticides: chlorpyrifos, alphacypermetrin, bromopropylate, carbendazim, and mancozeb
- Study the degradation processes of bromopropylate, coumaphos, chlordimeform, cymiazole, flumethrin, and fluvalinate in aqueous media by HPLC
- To develop a liquid-phase microextraction method using deep eutectic solvent for extraction and preconcentration of diazinon, metalaxyl, bromopropylate, oxadiazon, and fenazaquin pesticides from samples such as grape and sour cherry juices, fresh beet, cucumber, potato, and tomato followed by gas chromatography-flame ionisation detection
- To develop a multi-residue method for the determination of azinphos methyl, bromopropylate, chlorpyrifos, dimethoate, parathion methyl, and phosalone in apricots and peaches using negative chemical ionisation ion trap technology
- Simultaneous determination of amitraz, bromopropylate, coumaphos, cymiazole, and fluvalinate residues in honey by solid-phase extraction and GC-MS
Caution: P273 - P391 - P501