Supelco® TraceCERT® Organic Reference Standard, Procymidone
Procymidone, Certified reference material, TraceCERT®, 50 mg
Catalog # 75120-50MG
Supplier: Merck
Supelco® TraceCERT® Organic Reference Standard, Procymidone
Catalog # 75120-50MG
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Specifications
Specifications
- Pk:50 mg
- Standards:Certified reference material, TraceCERT®
- Description:Procymidone
- Cat. No.:75120-50MG
- M-Clarity quality level:MQ300
- Storage conditions:15...25 °C
- MDL No.:MFCD00078740
- Molecular weight:284,14
- Shipping conditions:STD
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



