Order Entry
Switzerland
ContactUsLinkComponent
Sodium methoxide 0,5 M for gas chromatography, Sigma-Aldrich®
Flammable
Toxic
Corrosive
Health
Sodium methoxide 0,5 M for gas chromatography, Sigma-Aldrich®
Catalog # 92446-5ML
Supplier:  Merck
New Product
CAS Number:  
Flammable
Toxic
Corrosive
Health
Sodium methoxide 0,5 M for gas chromatography, Sigma-Aldrich®
Catalog # 92446-5ML
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance (color):
    Colorless
  • Appearance (form):
    Liquid
  • Titration (molarity):
    Titration with hcl
  • Remarks on GC:
    Suitibility for gc derivatization complying
  • Density D20/4:
    0.805 - 0.817
  • Infrared spectrum:
    Conforms to structure

Specification Test Results

About this item

 :
 :54.02 g/mol
Density:0,805…0,817 g/ml (20 °C)
ADR:3,II
 :M-Clarity™ quality level = MQ100
UN:1289
 :GHS02
 :GHS06
 :GHS05
 :GHS08
CAS Number:124-41-4
Formula:CH₃NaO
MDL Number:MFCD00012179