SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
MycoLight™ PMA [Propidium Monoazide]
MycoLight™ PMA [Propidium Monoazide]
MycoLight™ PMA [Propidium Monoazide]
MycoLight™ PMA [Propidium Monoazide]
Catalog # AATB24215
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
MycoLight™ PMA [Propidium Monoazide]
Catalog # AATB24215
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
Product Details