Skip to Header
Skip to Main Content
Skip to Footer
Erweiterte Suche
Erweiterte Suche
Sicherheitsdatenblatt (SDS)
Zertifikate
Herstellerdaten
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
mFluor™ Violet 610-streptavidin conjugate
mFluor™ Violet 610-streptavidin conjugate conjugate
mFluor™ Violet 610-streptavidin conjugate conjugate
mFluor™ Violet 610-streptavidin conjugate conjugate
Catalog # AATB47272
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
mFluor™ Violet 610-streptavidin conjugate conjugate
Catalog # AATB47272
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Pk:
100 µG
Seq:
1286
Specifications
About this item
Produktdetails
:
Produktdetails