Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
mFluor™ Blue 660 tyramide
mFluor™ Blue 660 tyramide
mFluor™ Blue 660 tyramide
Catalog # AATB11051
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Bild vergrößern
mFluor™ Blue 660 tyramide
Catalog # AATB11051
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
200
Seq:
0200
Specifications
About this item
Product Details
:
:
770 g/mol
Product Details