Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(and-6)-Carboxy SNARF-1
5-(and-6)-Carboxy SNARF-1
5-(and-6)-Carboxy SNARF-1
5-(and-6)-Carboxy SNARF-1
Catalog # AATB21226
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
5-(and-6)-Carboxy SNARF-1
Catalog # AATB21226
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
453.45 g/mol
Product Details