Anti-TMEM16A Rabbit Polyclonal Antibody
ANTIA100737-100
New Product
- Antibody type:Primary
- Antigen name:ANO 1
- Antigen symbol:TMEM16A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q5XXA6
- Antigen synonyms:DOG 1|Discovered on gastrointestinal stromal tumors protein 1|Anoctamin1|Anoctamin 1 calcium activated chloride channel|Anoctamin 1|Anoctamin-1|Ca2+ activated Cl- channel|ANO1|ANO1_HUMAN
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:114 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human TM16A (amino acids 921 - 970)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to TM16A for IF and ELISA with samples derived from human.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: TMEM16A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human