Anti-Solute carrier family 22 member 5 Rabbit Polyclonal Antibody
Catalog # ANTIA100112-100
Anbieter: ANTIBODIES.COM
New Product
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:CDSP
- Antigen-Symbol:Solute carrier family 22 member 5
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# O76082
- Antigen-Synonyme:High-affinity sodium-dependent carnitine cotransporter|Solute carrier family 22 member 5|OCTN2|Solute carrier family 22 (organic cation/carnitine transporter) member 5|Organic cation/carnitine transporter 2|OCTN2VT|Slc22a5|S22A5_HUMAN
- Lagerungspuffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:62 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transporttemperatur:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SLC22A5 (amino acids 300 - 349)
- Reinigung:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Packung:Plastic vial
- VE:100 µG
Technische Daten
Über diesen Artikel
Rabbit polyclonal antibody to SLC22A5 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Solute carrier family 22 member 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human