Anti-HORMAD1 Rabbit Polyclonal Antibody
Catalog # ANTIA100163-100
Anbieter: ANTIBODIES.COM
New Product
Technische Daten
- Antibody type:Primary
- Antigen name:Cancer/testis antigen CT46
- Antigen symbol:HORMAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q86X24
- Antigen synonyms:Testis tissue sperm binding protein Li 86P|HORMA domain containing protein 1|OTTHUMP00000032933|HORMA domain containing 1|NOHMA|HORMAD1|HORMA domain containing protein|Newborn ovary HORMA protein|RP11-363I22 1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:45 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human HORMAD1 (amino acids 321-370)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Technische Daten
Über diesen Artikel
Rabbit polyclonal antibody to HORMAD1 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:5000
Type: Primary
Antigen: HORMAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human