Anti-Desmin Mouse Recombinant Antibody [Clone: rDES/1711]
Codice catalogo ANTIA278471-100
Nome del fornitore: ANTIBODIES.COM
New Product
Specifiche
- Antibody type:Primary
- Antigen name:CMD1I
- Antigen symbol:Desmin
- Clonality:Recombinant
- Clone:rDES/1711
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Hamster,Chicken,Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# P17661
- Antigen synonyms:CSM1|FLJ41013|DESM_HUMAN|DES|Desmin|FLJ12025|FLJ39719|FLJ41793|CSM2
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:52 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human Desmin protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifiche
Informazioni su questo articolo
Recombinant mouse monoclonal [rDES/1711] antibody to Desmin for IHC-P with samples derived from human, mouse, rat, hamster and chicken.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Desmin
Clonality: Recombinant
Clone: rDES/1711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Hamster, Chicken