Anti-Lambda Light Chain Rabbit Recombinant Antibody [clone: LLC/3774R]
Plastic vial
Catalog # ANTIA278001-100
Supplier: Antibodies.com
Anti-Lambda Light Chain Rabbit Recombinant Antibody [clone: LLC/3774R]
Catalog # ANTIA278001-100
Supplier: Antibodies.com
Jump to:
Product Details & Documents
Recombinant rabbit monoclonal [LLC/3774R] antibody to Lambda Light Chain for IHC-P with samples derived from Human.
Recommended dilutions: IHC-P: 1 to 2 µg/ml
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Type: Primary
Antigen: Lambda Light Chain
Clonality: Recombinant
Clone: LLC/3774R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Seq:1286
- Pack type:Plastic vial
- Clonality:Recombinant
- Purification:Protein A/G chromatography
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Concentration:200 µg/ml
- Molecular weight:~22,5 kDa
- Shipping temperature:Shipped on blue ice at 4 °C
- Antigen name:Bence Jones protein
- Cat. No.:ANTIA278001-100
- Reactivity:Human
- Gene ID:UniprotID# P01701, P01842
- Antigen synonyms:Ig lambda chain V-I region BL2|Ig lambda chain V-I region EPS|Ig lambda chain V-I region NEW|Ig lambda chain V-I region NIG-64|IGLV1-51|Immunoglobulin lambda variable 1-51
- Immunogen:Recombinant full-length human Lambda Light Chain
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Tested applications:IHC-P
- Antigen symbol:Lambda Light Chain
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Clone:LLC/3774R
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



