Order Entry
Switzerland
ContactUsLinkComponent
Anti-Thioredoxin/TRX Rabbit Polyclonal Antibody
Anti-Thioredoxin/TRX Rabbit Polyclonal Antibody
N° de catalogue ANTIA95540-100
Fournisseur:  ANTIBODIES.COM
New Product
undefined
Anti-Thioredoxin/TRX Rabbit Polyclonal Antibody
N° de catalogue ANTIA95540-100
Fournisseur:  ANTIBODIES.COM
New Product

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    ADF
  • Antigen symbol:
    Thioredoxin/TRX
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P10599
  • Antigen synonyms:
    Thioredoxin|Surface-associated sulphydryl protein|DKFZp686B1993|testicular tissue protein Li 199|MGC61975|ATL derived factor|ATL-derived factor|Surface associated sulphydryl protein|SASP
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    11 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human Thioredoxin (amino acids 49-98)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Spécifications

À propos de cet article

Rabbit polyclonal antibody to Thioredoxin for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Thioredoxin/TRX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat