Anti-ATF1 Rabbit Polyclonal Antibody
Catalog # ANTIA95037-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Activating transcription factor 1
- Antigen symbol:ATF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P18846
- Antigen synonyms:cAMP dependent transcription factor 1|ATF1_HUMAN|ATF1 FUS fusion gene|ATF1|cAMP-dependent transcription factor ATF-1|ATF1 EWS fusion gene|Cyclic AMP dependent transcription factor ATF1|Cyclic AMP dependent transcription factor ATF 1|ATF 1
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:29 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic phospho-peptide derived from human ATF1 around the phosphorylation site of Ser63
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising phospho peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to ATF1 (phospho Ser63) for WB and ELISA with samples derived from Human and Mouse.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: ATF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse