Anti-STING Rabbit Polyclonal Antibody
Catalog # ANTIA309483-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:endoplasmic reticulum IFN stimulator
- Antigen symbol:STING
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q3TBT3
- Antigen synonyms:MPYS|N terminal methionine proline tyrosine serine plasma membrane tetraspanner|hSTING|Endoplasmic reticulum interferon stimulator|ERIS|Mediator of IRF3 activation|Mitochondrial mediator of IRF3 activation|hMITA|FLJ38577
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% thiomersal
- Molecular weight:33 kDa/35 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of Mouse STING.
- Tested applications:IHC
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to STING for WB and IHC with samples derived from human, mouse and rat.
- Validated applications: WB, IHC
- Recommended dilutions: WB: 1:500 to 1:1,000, IHC: 1:50 to 1:200
Type: Primary
Antigen: STING
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat