Anti-AMPD1 Rabbit Polyclonal Antibody
N° de catalogue ANTIA97051-100
Fournisseur: ANTIBODIES.COM
New Product
Spécifications
- Antibody type:Primary
- Antigen name:Adenosine monophosphate deaminase 1
- Antigen symbol:AMPD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# P23109
- Antigen synonyms:AMPD|Ampd01|Adenosine monophosphate deaminase 1 (isoform M)|MAD|AMP deaminase isoform M|AMPD1|AMPD 1|AMP deaminase 1|MADA
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:86 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human AMPD1 (amino acids 261-310)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Spécifications
À propos de cet article
Rabbit polyclonal antibody to AMPD1 for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:40000
Type: Primary
Antigen: AMPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat