Anti-KCNT1 Rabbit Polyclonal Antibody
Katalognr. ANTIA97019-100
Anbieter: ANTIBODIES.COM
New Product
Technische Daten
- Antibody type:Primary
- Antigen name:bA100C15.2
- Antigen symbol:KCNT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# Q5JUK3
- Antigen synonyms:KCa4.1|KCNT1_HUMAN|Potassium channel, subfamily T, member 1|Potassium channel, sodium activated subfamily T, member 1|ENFL5|EIEE14|Sequence like a calcium-activated K+ channel|Potassium channel subfamily T member 1|KCNT1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:138 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human KCNT1 (amino acids 1019-1068)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Technische Daten
Über diesen Artikel
Rabbit polyclonal antibody to KCNT1 for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: KCNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat