Anti-NUAK1 Goat Polyclonal Antibody
Catalog # ANTIA84498-100
Anbieter: ANTIBODIES.COM
New Product
Technische Daten
- Antibody type:Primary
- Antigen name:AMP activated protein kinase family member 5
- Antigen symbol:NUAK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:Omphalocele kinase 1|Nuak1|NUAK family SNF1 like kinase|AMPK related protein kinase 5|ARK5|AMPK-related protein kinase 5|NUAK family SNF1-like kinase 1|NUAK1_HUMAN|Probable serine/threonine protein kinase KIAA0537
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-QNRPRPQYLKRYRNR
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human NUAK1 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Technische Daten
Über diesen Artikel
Goat polyclonal antibody to NUAK1 for ELISA, IF and flow cytometry with samples derived from human.
- Validated applications: ELISA, IF, Flow Cytometr
Type: Primary
Antigen: NUAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human