Order Entry
Switzerland
ContactUsLinkComponent
Anti-LXN Goat Polyclonal Antibody (Biotin)
Anti-LXN Goat Polyclonal Antibody (Biotin)
Catalog # ANTIA84314-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-LXN Goat Polyclonal Antibody (Biotin)
Catalog # ANTIA84314-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ECI
  • Antigen symbol:
    LXN
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    Protein MUM|LXN_HUMAN|Endogenous carboxypeptidase inhibitor|Tissue carboxypeptidase inhibitor|Latexin protein|TCI|LXN|MUM|Latexin
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    C-QYGTKVKHNSRLPKE
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to Human LXN (C terminal)
  • Tested applications:
    IHC
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Goat polyclonal antibody to LXN (Biotin) for ELISA, WB, IHC and IF with samples derived from human, mouse, rat and canine.

  • Validated applications: ELISA, WB, IHC, I

Type: Primary
Antigen: LXN
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat;Dog